Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 92743PU HyperMax™ Gloves, Polyurethane Coating

Catalog Number p-7138423
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless salt and pepper colored HPPE/synthetic shell that offers cut, tear and abrasion resistance. Gray polyurethane coated palm and fingertips provide a durable grip and hides dirt, grit and grime.

  • 13-gauge seamless salt & pepper colored HPPE/synthetic shell
  • Gray PU palm and fingers
  • Comfortable and easy to wear
  • Long-lasting dexterity
ANSI Abrasion Rating ANSI 5
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A5
Material HPPE
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Automotive Assembly, Material Handling, Landscaping, Glass Handling
Glove Cuff Style Knit
Hem Color Multi-colored
Model 92743PUS
Series Cut Pro™ 92743PU
Yarn Gauge 13
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit