Learn More
BD™ MAX™ UVE Sample Buffer Tube
BD™ MAX™ UVE Sample Buffer Tube for use with UVE Specimen Collection Kits
Supplier:  BD™ 443420
Catalog No. B443420
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
View alternate product
This item is not returnable. View return policy
Description
Description
  • Buffer contains approximately 1.5 mL of potassium phosphate, ethylenediaminetetraacetic acid (EDTA), Tween™ 20 and ProClin™
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UVE Sample Buffer Tube
48/Pk.