Learn More
Diversified Spaces™ Maple Table with Plain Apron and Solid Epoxy Top: Leg Brace and Stretcher Option
SCI_ED
Solid Epoxy tops are impervious to normal laboratory chemicals and heat. Diversified Spaces™ Maple Tables with Epoxy Tops are ideal for high school and above with gas and other heat producing processes.
$1176.00 - $1757.00
Specifications
Adjustable No
Certifications/Compliance SEFA compliant and MAS Certified Green™
Finish Chemical resistant, earth-friendly UV
For Use With (Application) High school and above with gas and other heat producing processes
Includes Leg Brace and Stretcher
View More Specs
Products 15
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Height (English) Includes Price Quantity & Availability  
S41963
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7306M30S
24 x 72 in. (60.96 x 182.88 cm) 30 in. Leg Brace and Stretcher
Each for $1,507.00
Add to cart
S41973
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7306M36S
24 x 72 in. (60.96 x 182.88 cm) 36 in. Leg Brace and Stretcher
Each for $1,580.00
Add to cart
S42174
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7606M30S
24 x 60 in. (60.96 x 152.4 cm) 30 in. Leg Brace and Stretcher
Each for $1,322.00
Add to cart
S41970
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7306M34S
24 x 72 in. (60.96 x 182.88 cm) 34 in. Leg Brace and Stretcher
Each for $1,580.00
Add to cart
S47514
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7106M36S
24 x 48 in. (60.96 x 121.92 cm) 36 in. Leg Brace and Stretcher
Each for $1,250.00
Add to cart
S42774
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7146M34S
30 x 60 in. (76.2 x 152.4 cm) 34 in. Leg Brace and Stretcher
Each for $1,627.00
Add to cart
S42188
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7606M36S
24 x 60 in. (60.96 x 152.4 cm) 36 in. Leg Brace and Stretcher
Each for $1,396.00
Add to cart
S47505
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7106M30S
24 x 48 in. (60.96 x 121.92 cm) 30 in. Leg Brace and Stretcher
Each for $1,176.00
Add to cart
S47874
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7146M30S
30 x 60 in. (76.2 x 152.4 cm) 30 in. Leg Brace and Stretcher
Each for $1,553.00
Add to cart
S47888
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7146M36S
30 x 60 in. (76.2 x 152.4 cm) 36 in. Leg Brace and Stretcher
Each for $1,627.00
Add to cart
S48009
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7156M30S
30 x 72 in. 30 in. Leg Brace and Stretcher
Each for $1,684.00
Add to cart
S48017
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7156M36S
30 x 72 in. 36 in. Leg Brace and Stretcher
Each for $1,757.00
Add to cart
S48503
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7206M34S
24 x 54 in. (60.96 x 137.16 cm) 34 in. Leg Brace and Stretcher
Each for $1,283.00
Add to cart
S48496
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7206M30S
24 x 54 in. (60.96 x 137.16 cm) 30 in. Leg Brace and Stretcher
Each for $1,209.00
Add to cart
S48510
SCI_ED
missing translation for 'viewDocuments'
Diversified Spaces™
P7206M36S
24 x 54 in. (60.96 x 137.16 cm) 36 in. Leg Brace and Stretcher
Each for $1,283.00
Add to cart
Description
Description
  • 1 in. Epoxy Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes leg braces and stretcher
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Maple Construction
Specifications
Specifications
No
Chemical resistant, earth-friendly UV
Leg Brace and Stretcher
226.79 kg
Solid Epoxy
Maple
SEFA compliant and MAS Certified Green™
High school and above with gas and other heat producing processes
500 lb.
Plain Apron
Plain Apron Table
SDS
Documents
Documents
Product Certifications
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'