Learn More
Diversified Spaces™ Maple Table with Plain Apron and Epoxy Resin Top
With non-emitting, chemical-resistant UV finish.
Specifications
Certifications/Compliance | SEFA compliant and MAS Certified Green |
---|---|
Finish | UV |
Includes | Nuts and washers, leg boots and levelers. |
Load Bearing Capacity (English) | 500 lb. |
Load Bearing Capacity (Metric) | 226.79 kg |
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Price | Quantity & Availability | ||||
S41962
|
Diversified Spaces™
P7306M30N |
24 x 72 in. | 30 in. |
Each for $1,431.00
|
N/A | ||||
S41969
|
Diversified Spaces™
P7306M34N |
24 x 72 in. | 34 in. |
Each for $1,505.00
|
N/A | ||||
S42173
|
Diversified Spaces™
P7606M30N |
24 x 60 in. | 30 in. |
Each for $1,247.00
|
N/A | ||||
S42180
|
Diversified Spaces™
P7606M34N |
24 x 60 in. | 34 in. |
Each for $1,322.00
|
N/A | ||||
S42770
|
Diversified Spaces™
P7146M30N |
30 x 60 in. | 30 in. |
Each for $1,477.00
|
N/A | ||||
S47513
|
Diversified Spaces™
P7106M36N |
24 x 48 in. | 36 in. |
Each for $1,176.00
|
N/A | ||||
S47504
|
Diversified Spaces™
P7106M30N |
24 x 48 in. | 30 in. |
Each for $1,101.00
|
N/A | ||||
S47880
|
Diversified Spaces™
P7146M34N |
30 x 60 in. | 34 in. |
Each for $1,552.00
|
N/A | ||||
S48008
|
Diversified Spaces™
P7156M30N |
30 x 72 in. | 30 in. |
Each for $1,608.00
|
N/A | ||||
S48509
|
Diversified Spaces™
P7206M36N |
24 x 54 in. | 36 in. |
Each for $1,208.00
|
N/A | ||||
S48502
|
Diversified Spaces™
P7206M34N |
24 x 54 in. | 34 in. |
Each for $1,208.00
|
N/A | ||||
Description
- Constructed from maple
- Plain apron with heavy-duty 13-gauge steel corner braces
- Epoxy resin table surface
Specifications
SEFA compliant and MAS Certified Green | |
Nuts and washers, leg boots and levelers. | |
226.79 kg | |
Plain Apron Table |
UV | |
500 lb. | |
1 in. Epoxy Resin top | |
Maple |