Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mapa Pioneer Mapa™ Professional Krytech Cut Protective Sleeves
Click to view available options
Cut Protective Sleeves; Length: 600mm (24 in.)
Description
- Sleeves with adjustable velcro strap
- Cut protective cuff made from HDPE
- Attaches at top of arm for comfort and safety
- Sleeve cuff does not slip and keeps arm safe
- Performances of sleeves are guaranteed up to 5 cleaning cycles
Specifications
Length (English) | 24 in. |
Length (Metric) | 600 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction