Learn More
Malvern Panalytical 100 nm / 15 mL Polystyrene Standard
Supplier: Malvern Panalytical LTX3100A
15 ml Latex Standard with a Certified Diameter of 100nm.Nanosphere Size Standards are highly uniform polystyrene spheres calibrated by NIST traceable standards and packaged as aqueous suspensions in dropper-tipped bottles. 1% solids, optimized for easy dispersal and colloidal stability. Includes Certificate of Calibration and Traceability to NIST.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.