Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Maleic Acid
SCI_ED

Catalog Number s25415
Click to view available options
Quantity:
200 g
250 g
Packaging:
Solid
110-16-7
C4H4O4
116.07
MFCD00063177
VZCYOOQTPOCHFL-OWOJBTEDSA-N
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
444266
CHEBI:18300
(2E)-but-2-enedioic acid
OC(=O)\C=C\C(O)=O
110-16-7
C4H4O4
116.07
MFCD00063177
VZCYOOQTPOCHFL-OWOJBTEDSA-N
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
444266
CHEBI:18300
(2E)-but-2-enedioic acid
OC(=O)\C=C\C(O)=O

This product is for educational use - no C of A is available.

Color White
Assay Percent Range >98%
Linear Formula HO2CCH:CHCO2H
Formula Weight 116.07
Density 1.59g/mL
Quantity 200 g
Packaging Solid
Chemical Name or Material Maleic Acid
ShelfLife 5 years
DOTInformation DOT Class 8, III, : Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit