Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carson™ MagniRama 3x Power 50 mm Assorted Color Glass Magnifying Glasses
Pack of 50 magnifying glasses. These are 3x power, 50 mm all-glass magnifiers that provide crystal-clear optics. These are great learning tools for children of all ages. They are small enough and lightweight enough to fit in backpacks or even pockets.
Supplier: Carson™ JD3MU
This item is not returnable.
View return policy
Description
- The MagniRama 3x Power 50 mm Magnifying Glasses - Pack of 50
- These 3x power, all-glass magnifiers provide crystal-clear optics and are a great learning tool for children of all ages.
- They are small enough and lightweight enough to fit in backpacks or even pockets.
- Great for the classroom, goody bags, exploring science, nature and so much more.
- They are backed by Carson's limited lifetime warranty.
Specifications
Magnifying Glass | |
Inspection | |
1.97 in. | |
Glass |
3x | |
50 Pk. | |
50 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction