Learn More
V&P Scientific Inc MAGNETIC PLATE, 384 Well, Standard, Flat Bottom, 425 Round Posts, BioTek Plate Washer Compatible
Supplier: V&P Scientific Inc VP 771BTG42AZ
MAGNETIC BEAD SEPARATION PLATE, 384 Well, Standard Plate, Flat Bottom, 425 Round Posts, 52 MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, 4 Sided Pellet Location, 3.1mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, BioTek Plate Washer Compatible, Non-SLAS Footprint, No Registration Base