Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Magnesium Carbonate, Light Powder, USP, 40-43.5%, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
500 g
6 x 500 g
12 kg
4 x 2.5 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Magnesium Carbonate, Light Powder, USP is used in anti-acid preparations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Quantity | 100 g |
Percent Purity | 40 to 43.5% |
Grade | USP |
Packaging | Poly Bottle |