Learn More
V&P Scientific Inc MAGNETIC PLATE, 96 Well, Standard or Deep Well, Round or Conical Bottom, 13 Bars, 50MGO, 2 Sided Pellet Location
Supplier: V&P Scientific Inc VP 771MDWZM1
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 13 Bars, 50MGO, NdFeB Magnets, White Polycarbonate Magnet Frame, 2 Sided Pellet Location, Bar 3mm Wide, Axial - North Poles Up Magnetic Orientation, SLAS Footprint, Includes Registration Base
This item is not returnable.
View return policy