Learn More
METTLER TOLEDO Transmitter M100 SM 1-Wire
Supplier: METTLER TOLEDO 30365366
The M100 Sensor Mount (SM) is a single-channel transmitter for connecting 1-wire ISM pH, DO (amperometric) and CO2 sensors to a biocontroller. The Bluetooth 4.0 interface is compatible with the PC-based and mobile versions of iSense software.