Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Workspace Wide Storage Cabinets
SDPSCI_ED

Double door cabinet with adjustable shelves.

Supplier:  Lyon LLC LMF1001

Catalog No. S510221MF

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.


Description

Description

  • With “Quiet door” feature
  • Available shelf reinforcements boost load capacity
  • Four shelves adjustable every 2 in. (extra shelves are available)

Order Info

Dimension: 21D x 36W x 78 in. H
Capacity/Shelf: 180 lb. (81.6kg)

Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit