Learn More
MP Biomedicals™ LymphoSpinner™ Lymphocyte Isolation Tubes
An innovative separation tube with a porous membrane for optimal separation of lymphocytes and mononuclear cells from blood and tissues.
$214.00 - $275.00
Catalog Number | Mfr. No. | Volume (Metric) Working | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) Working | Price | Quantity & Availability | |||||
09-169-2425
|
MP Biomedicals™
091692425 |
15 mL |
Each for $214.00
|
|
|||||
09-169-2525
|
MP Biomedicals™
091692525 |
50 mL |
Each for $275.00
|
|
|||||
Description
- Biologically inert barrier prevents mixture of the sample with the Lymphocyte Separation Medium (LSM™)
- Maintains discontinuous gradient and improves separations with higher yield and less processing time
- Sterile and designed for single use only
Specifications
Isolation Tube | |
25 Tubes | |
Sterile |