Learn More
Lyle and Louise: Stained with Suspicion™ Blood Detection and Evidence Processing

Introduces students to presumptive blood tests and evidence processing.
$62.40 - $164.30
Specifications
Grade | 9 to Undergraduate |
---|---|
For Use With (Application) | For introduces students to presumptive blood tests and evidence processing |
Includes | Six bottles of Hydrogen Peroxide, six bottles of Phenolphthalein solution, six sets of Cotton swabs, six sets of positive controls and presumptive test cards, six carpet scraps from the crime scene, fully illustrated teacher's manual with technique background |
Requires | Lab gloves, distilled water, rulers for each group, camera, tweezers |
Class Size | 30 Students |
Includes: Six bottles of hydrogen peroxide, six bottles of phenolphthalein solution, six sets of cotton swabs, six sets of positive controls and presumptive test cards, six carpet scraps from the crime scene, fully illustrated teacher's manual with technique background
Requires: Lab gloves, distilled water, rulers for each group, camera (optional), tweezers (optional)
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S02173
![]() |
Crosscutting Concepts LLC
VXHLLBDD |
Mystery Of Lyle And Louise: Stained With Suspicion™ Blood Detection And Evidence Processing Forensic Science Curriculum Kit |
Each for $164.30
|
|
S02173R
![]() |
Crosscutting Concepts LLC
VXHLLBDDRF |
Mystery Of Lyle And Louise: Stained With Suspicion™ Blood Detection And Evidence Processing Forensic Science Refill Kit |
Each for $62.40
|
|
Description
Helps determine if a substance at the crime scene is blood and teaches forensic techniques involved in evidence processing.
- Analyze different items to find traces of blood
- Process the piece of evidence as processed at the actual crime scene
Order Info
For up to 30 students working in six groups. Kit is geared toward high school and undergraduate courses and matched to National Science Education Standards.
Specifications
9 to Undergraduate | |
Six bottles of Hydrogen Peroxide, six bottles of Phenolphthalein solution, six sets of Cotton swabs, six sets of positive controls and presumptive test cards, six carpet scraps from the crime scene, fully illustrated teacher's manual with technique background | |
30 Students |
For introduces students to presumptive blood tests and evidence processing | |
Lab gloves, distilled water, rulers for each group, camera, tweezers |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.