Learn More
Lyle and Louise: Shoe Must Fit™ Footprint Analysis

Using a shoe print collected at the crime scene, students will be able to determine critical information about the fictional criminal by analyzing and comparing shoe castings.
$47.60 - $164.30
Specifications
Grade | 9 to Undergraduate |
---|---|
Content And Storage | Room temperature |
Class Size | 30 Students |
Includes: Six true-to-size castings of the evidence footprint for each group, six photos of additional shoe print taken from crime scene, six bags of dental stone (stronger and hardens more quickly than plaster) for each group, six sets of shoe data records from SoleMate (database for forensic identification of shoe prints from Foster + Freeman), fully illustrated teacher's manual with technique background
Requires: Yard or meter stick for each group, water, computer with internet access for each group
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Includes | Requires | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Includes | Requires | Price | Quantity & Availability | ||
S02170
![]() |
Crosscutting Concepts LLC
VXHLLFTP |
For teaching determination of critical information about suspect using shoe print | Mystery Of Lyle And Louise: Shoe Must Fit™ Footprint Analysis Forensic Science Curriculum Kit | Fully illustrated teacher's manual with technique background, 6 true to size castings of the evidence footprint for each group, 6 photos of additional shoe print taken from crime scene, 6 bags of dental stone, 6 sets of shoe data records from solemate | Yard stick or meter stick for each group, water, computer with internet access for each group |
Each for $164.30
|
|
S02170R
![]() |
Crosscutting Concepts LLC
VXHLLFTPRF |
For footprint analysis | Mystery Of Lyle And Louise: Shoe Must Fit™ Footprint Analysis Forensic Science Refill Kit | 6 true-to-size castings of the evidence footprint for each group, 6 photos of additional shoe print taken from crime scene, 6 bags of dental stone for each group, 6 sets of shoe data records from solemate, fully illustrated teacher's manual with technique background | Yard or meter stick for each group, water, computer with internet access for each group |
Each for $47.60
|
|
Description
- Students perform a lab to compare height with shoe size and stride length
- Students take castings of their own footprints and use both a casting of the evidence shoe print and a photograph of an additional shoe print found at the scene of the crime to determine individual characteristics about the shoeprints of a suspect or suspects
Order Info
Supplies are included for up to 30 students working in six groups. Kit is geared to high school and undergraduate courses and is matched toward National Science Education Standards.
Specifications
9 to Undergraduate | |
Room temperature | |
30 Students |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.