Learn More
Lyle and Louise: Read the Found Prints™ Fingerprint Analysis
For the identification of an individual's fingerprints.
Supplier: Crosscutting Concepts LLC VXHLLFGP
Includes: 30 hinge lifters, six brushes, dusting powder, professional fingerprinting ink pads, evidence fingerprint from office, fingerprint cards from the suspects, fully illustrated teacher's manual with technique background
Requires : Lab gloves, overhead transparencies or clear glass surface (such as an available window or glass from a picture frame), weigh boat, Petri dish or paper bowl for powder, magnifying glass (optional)
Description
Students analyze a fingerprint found at the business offices of Lyle Mondelo.
- Kit introduces students to the fingerprinting techniques used by the FBI and other police professionals
- Learn and perform both rolling and lifting prints
- Analyze a fingerprint
- Compare it to several sets of fingerprints to determine if there was suspicious activity
This kit is geared toward high school and undergraduate courses, and is matched to National Science Education Standards. It is endorsed by Sirchie Finger Print Laboratories, a comprehensive manufacturer and distributor of forensic evidence collection supplies, allowing your students to roll and lift prints with the materials used by real forensic scientists.
Order Info
For up to 30 students working in six groups.
Specifications
Mystery of Lyle and Louise: Read the Found Prints™ Fingerprint Analysis Curriculum Kit | |
For identification of individual's fingerprints | |
Lab gloves, overhead transparencies or clear glass surface, weigh boat, petri dish or paper bowl for powder, magnifying glass |
9 to Undergraduate | |
30 hinge lifters, six brushes, dusting powder, professional fingerprinting ink pads, evidence fingerprint from office, fingerprint cards from the suspects, fully illustrated teacher's manual with technique background | |
30 Students |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.