Learn More
Lyle and Louise: Prescription for Crime™ Drug Testing and Analysis
Helps students determine the identity of a white powder found in an abandoned truck.
Supplier: Crosscutting Concepts LLC VXHLLDRT
Includes: Six bottles of isopropyl alcohol, six bottles of Lugol's iodine solution, mystery powder for each group, six known powders for each group, spot plates for each group, powder dispenser spatulas for each group, fully illustrated teacher's manual with technique background
Requires : Lab gloves, distilled water, pipets for dispensing distilled water, permanent marker for labeling
Description
- Several presumptive tests are performed on the unknown white powder
- Following the presumptive tests, confirmatory drug tests are run in a police lab using gas chromatography-mass spectrometry
- Students receive the data for the confirmatory drug test
Order Info
For up to 30 students working in six groups. Kit is geared toward high school and undergraduate courses and matched to National Science Education Standards.
Specifications
9 to Undergraduate | |
Sem data for 12 particles (graphs and images), 6 positive and negative control swabs, gunshot residue swabs taken from 6 different surfaces, 50 alcohol swabs, pipettes for each group, 6 bottles of diphenylamine, 6 bottles of sodium rhodizonic acid, fully | |
Lab gloves, distilled water, pipets for dispensing distilled water, permanent marker for labeling | |
30 Students |
For teaching drug testing and analysis procedures | |
Mystery Of Lyle And Louise: Prescription For Crime™ Drug Testing And Analysis Curriculum Kit | |
Room temperature |