Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lyle and Louise: Patterns of Murder™ Blood Spatter Analysis

Blood spatter analysis provides more clues to the mystery.
$56.60 - $211.50
Specifications
Grade | 9 to Undergraduate |
---|---|
Includes | Six blood drop analysis stations, six blood spatter mockups, six protractors, six measuring tapes, six bottles of synthetic blood, six glass pipets with rubber bulbs, six packs of string, 24 sheets of spatter paper, box of pushpins, teaching manual |
Requires | Lab gloves, computer with MS Excel |
Class Size | 30 Students |
Includes: Six blood drop analysis stations, six blood spatter mockups, six protractors, six measuring tapes, six bottles of synthetic blood, six glass pipets with rubber bulbs, six packs of string, 24 sheets of spatter paper, box of pushpins, teaching manual
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
S94585
![]() |
Crosscutting Concepts LLC
VXH10133 |
For teaching blood splatter pattern analysis procedures | Lyle And Louise: Patterns Of Murder™ Blood Spatter Analysis Curriculum Kit |
Each for $211.50
|
|
S94585R
![]() |
Crosscutting Concepts LLC
VXH10285 |
For used to replenish consumable components | Lyle And Louise: Patterns Of Murder™ Blood Spatter Analysis Refill Kit |
Each for $56.60
|
|
Description
Students determine the relationship between the height and angle of a blood drop impact and the size and shape of the spatter.
- Synthetic blood is dropped onto paper at different heights and angles; the resulting spatter is measured and documented
- Data is analyzed and a mathematical model is built using Microsoft™ Excel™ or another spreadsheet program
- Students use blood spatter patterns to determine point of impact by using strings from the spatter to the crime scene walls
- Class size: six groups of up to five students per group
Specifications
9 to Undergraduate | |
Six blood drop analysis stations, six blood spatter mockups, six protractors, six measuring tapes, six bottles of synthetic blood, six glass pipets with rubber bulbs, six packs of string, 24 sheets of spatter paper, box of pushpins, teaching manual | |
Lab gloves, computer with MS Excel | |
30 Students |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title