Learn More
Lyle and Louise: Patterns of Murder™ Blood Spatter Analysis
Blood spatter analysis provides more clues to the mystery.
Supplier: Crosscutting Concepts LLC VXH10285
Includes: Six blood drop analysis stations, six blood spatter mockups, six protractors, six measuring tapes, six bottles of synthetic blood, six glass pipets with rubber bulbs, six packs of string, 24 sheets of spatter paper, box of pushpins, teaching manual
Description
Students determine the relationship between the height and angle of a blood drop impact and the size and shape of the spatter.
- Synthetic blood is dropped onto paper at different heights and angles; the resulting spatter is measured and documented
- Data is analyzed and a mathematical model is built using Microsoft™ Excel™ or another spreadsheet program
- Students use blood spatter patterns to determine point of impact by using strings from the spatter to the crime scene walls
- Class size: six groups of up to five students per group
Specifications
Lyle and Louise: Patterns of Murder™ Blood Spatter Analysis Refill Kit | |
For used to replenish consumable components | |
Lab gloves, computer with MS Excel |
9 to Undergraduate | |
Six blood drop analysis stations, six blood spatter mockups, six protractors, six measuring tapes, six bottles of synthetic blood, six glass pipets with rubber bulbs, six packs of string, 24 sheets of spatter paper, box of pushpins, teaching manual | |
30 Students |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.