Learn More
Lyle and Louise: Patterns of Murder™ Blood Spatter Analysis
SCI_ED
Blood spatter analysis provides more clues to the mystery.
Supplier:  Crosscutting Concepts LLC VXH10133
 View more versions of this product
Catalog No. S94585
Add to Cart
Includes: Six blood drop analysis stations, six blood spatter mockups, six protractors, six measuring tapes, six bottles of synthetic blood, six glass pipets with rubber bulbs, six packs of string, 24 sheets of spatter paper, box of pushpins, teaching manual
Description
Description
Students determine the relationship between the height and angle of a blood drop impact and the size and shape of the spatter.
  • Synthetic blood is dropped onto paper at different heights and angles; the resulting spatter is measured and documented
  • Data is analyzed and a mathematical model is built using Microsoft™ Excel™ or another spreadsheet program
  • Students use blood spatter patterns to determine point of impact by using strings from the spatter to the crime scene walls
  • Class size: six groups of up to five students per group
Specifications
Specifications
Lyle and Louise: Patterns of Murder™ Blood Spatter Analysis Curriculum Kit
For teaching blood splatter pattern analysis procedures
Lab gloves, computer with MS Excel
9 to Undergraduate
Six blood drop analysis stations, six blood spatter mockups, six protractors, six measuring tapes, six bottles of synthetic blood, six glass pipets with rubber bulbs, six packs of string, 24 sheets of spatter paper, box of pushpins, teaching manual
30 Students