Learn More
Lyle and Louise: Nature's Witness™ Forensic Entomology
SCI_ED
Learn how forensic entomologists use maggots to determine the post-mortem interval.
Supplier:  Crosscutting Concepts LLC VXH10135
Catalog No. S94587
Add to Cart
Includes: Six life stages of Species A (Lucilia sericata), six life stages of Species B (Bercaea haemorrhoidalis), six evidence collections, fully illustrated teacher's manual with technique background
Requires : Dissection microscope and tweezers
Description
Description
The bodies discovered in the cabin were badly decomposed and had large maggot masses on them. Students are introduced to the techniques of forensic entomology and use those techniques to examine insects collected at the scene.
  • Examine exemplars of the life stages of two fly species and develop a key for distinguishing between life stage and species
  • Use the key to identify a number of insects collected from the corpses
  • Record their information for the entire class to share
  • Compare the identification data to a life stage/species development chart and National Weather Service data to determine the post-mortem interval for both decedents
  • Class size: six groups of up to five students per group
Specifications
Specifications
9 to Undergraduate
Six life stages of species a (lucilia sericata), six life stages of species b (bercaea haemorrhoidalis), six evidence collections, fully illustrated teacher's manual with technique background
Entomology featherweight forceps, lab gloves, microscope or magnifying glass, computer with MS Excel
To learn how forensic entomologists use maggots to determine the post-mortem interval
Mystery Of Lyle And Louise: Nature'S Witness™ Forensic Entomology Forensic Science Curriculum Kit
30 Students