Learn More
Thermo Scientific™ Luminaris Color HiGreen qPCR Master Mix
Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.
$339.00 - $3772.00
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
FERK0393
|
Thermo Scientific™
K0393 |
1250 Reactions |
Each for $1,357.00
|
|
|||||
FERK0391
|
Thermo Scientific™
K0391 |
250 Reactions |
Each for $339.00
|
|
|||||
FERK0392
|
Thermo Scientific™
K0392 |
500 Reactions |
Each for $619.00
|
|
|||||
FERK0394
|
Thermo Scientific™
K0394 |
5000 Reactions |
Each for $3,772.00
|
|
|||||
Description
Thermo Scientific™ Luminaris Color HiGreen Master Mixes are ready-to-use qPCR master mixes with color and premixed UDG optimized for SYBR Green chemistry using standard cycling protocols.
- Blue master mix and Yellow Sample Buffer for easy pipetting
- UDG in the master mix to prevent carry-over contamination
- Equal performance of blue and colorless master mixes
- Versions with premixed ROX or fluorescein for different real-time platforms
- Standard cycling protocols
- Excellent qPCR performance
- Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity: single copy detection
- Wide linear range: accurate quantification across 9 orders of magnitude
- Reproducibility and convenience: ready-to-use 2X master mix
Specifications
SYBR | |
qPCR | |
Standard | |
Luminaris | |
Gene Expression | |
2 X |
High | |
Taq DNA Polymerase | |
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene | |
qPCR Master Mix | |
2X | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.