Learn More
Carson™ JK Series Close Focus Waterproof Binoculars



Superior close focus capabilities allow users to focus on objects as close as 6 feet. Carson JK Series Close Focus Waterproof Binoculars are ideal for wildlife viewing because it allows one to view the details of a bird or animal.
Supplier: Carson™ JK042
Includes: Pouch, neck strap, lens covers and lens cloth
Description
- A 10x42mm close focus roof prism binocular
- Features BAK-4 prisms and fully multi-coated optics for crisp, clear viewing
- Large 42mm objective lenses gather plenty of light to ensure the viewing image is bright even in low light conditions
- O-ring sealed and nitrogen purged making them waterproof and fogproof
- Twist-up eyecups stop in three positions to provide optimal eye relief for all users
- Large focus wheel turns smoothly for precise focus adjustments
- Independent right diopter adjustment makes it possible to adjust the focus to allow for strength differences in each eye
Specifications
10x | |
BAK-4 Prism | |
Hunting, birding, hiking or fishing. | |
1.4 lb. |
Fully Multi-coated | |
42 mm | |
Pouch, neck strap, lens covers and lens cloth |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.