Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Walter Products Lumbar Spinal Column
This replica of the lumbar spinal column consists of the 5 lumbar vertebrae, sacrum and coccyx. The intevertebral discs, spinal cord and lumbar nerves are also shown, as well as a prolapsed disc between the L3 and L4 vertebrae. Comes mounted on a stand.
Supplier: Walter Products B10262
Description
- Flexible and articulated lumbar spine
- Sacrum along with spinal cord and paired spinal nerves
- The intervertebral disc between L4 and L5 is herniated
- Mounted on a stand
Specifications
Model, Skeleton, Lumbar | |
30 cm | |
11 cm | |
1 | |
11 cm |
Education | |
Lumbar model & stand | |
PVC | |
0.82 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction