Learn More
Vidan™ Diagnostics Lucidplus™ BHB Plus Glucose Monitoring System
Lucidplus™ is a hand-held, quantiative monitoring system that measures Beta-Hydroxybutyrate, (BHB) and Glucose in whole blood. It is inteded for multi-patient use in the professional healthcare setting.
Supplier:  Vidan™ Diagnostics 300300
Catalog No. 23-300-101
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Measures Beta-hydroxybutyrate (BHB) and Glucose
  • Whole blood sample, capillary or venous
  • Quantitative results within 10 seconds
  • Small sample size, approximately 0.8 μL
  • Stores up to 1,000 results
  • Power Supply: one AAA battery
  • Drop ship from Vidan Diagnostics
  • Certifications: FDA 510(k), CLIA Waived
  • 2 year warranty
Specifications
Specifications
(1) Lucidplus BHB Plus Glucose Monitoring System, (1) Quick Guide, (1) Operator's manual
4.3 cm
0.13 kg
16.0 cm
1000 Tests