Learn More
Electron Microscopy Sciences Large Paraffin Dispenser, 50/60Hz, 4.5 Amp
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Electron Microscopy Sciences 6284010
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Holds 6.25 gallons of paraffin wax with clear lexan lid permits easy viewing of paraffin without contamination. High spigot accommodates large transfer vessels. The unit comes with microprocessor temperature regulation, built-in clock. Ambient to 65 degree C.The Paraffin Dispenser have been tested by a registered independent body to rigorous international quality and safety standards, and, have been found to be in compliance with UL, CSA and CE.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.