Learn More
MilliporeSigma™ Supelco™ LpDNPH Rezorian Cartridge, Bed wt. 350 mg
Solvent-compatible due to its polypropylene components and polyethylene frits
$217.00 - $1032.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-0235
|
MilliporeSigma™ Supelco™
54074U |
10/Pk. |
Each for $217.00
|
|
|||||
11-100-7703
|
MilliporeSigma™ Supelco™
54075U |
50/Pk. |
Each for $1,032.00
|
|
|||||
Description
LpDNPH Rezorian™ Cartridge are solvent-compatible due to its polypropylene components and polyethylene frits. It can be commonly used, either individually or connected in series. It is employed in-line with peristaltic pumps and low pressure HPLC systems, or with a syringe and a single sample processor.Specifications
Bed wt. 350 mg | |
3 mL |