Learn More
Sartorius Low Retention SafetySpace™ Filter Tips
Offers doubled security for sample recovery and improves pipetting precision
Catalog Number | Mfr. No. | Volume (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
S35896
![]() |
Sartorius
LHLF790011/EMD |
1 to 10 μL | 32 mm | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
S35897
![]() |
Sartorius
LHLF790101/EMD |
2 to 120 μL | 51 mm | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Low Retention Tips have been developed to acquire highly liquid-repellent surface by using a modern patented technology. This method produces tips with complete hydrophobic coverage, and with high chemical resistance.
- Hydrophobic tip surface combined with safety air gap between sample and filter improved sample recovery in two ways: through better liquid flow, and by reducing risk of sample penetrating filter
- Used for liquids such as detergents and solvents with low surface tension
- Prevents cross-contamination
- Made of recylcable polypropylene
- Suitable where multi-dispensing with excess liquid volume is used
- Liquid-repellent tip surface with chemical resistant
- Chemically resistant and non-coated tip would not interfere with sample
- Filtered and pre-sterilised
- DNase, RNase, endotoxin free
PCR/real-time PCR, Cloning, Sequencing, Other DNA and RNA techniques, SDS-PAGE and other protein analysis methods
Specifications
Filtered | |
Sterile |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.