Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ ProSieve™ Color Protein Markers
Visible markers for use in SDS-PAGE gels
$148.80 - $518.50
Specifications
For Use With (Application) | Monitoring protein separation prior to staining and provide confirmation of protein transfer in Western blotting |
---|
Catalog Number | Mfr. No. | Quantity | Formulation | Range | Storage Requirements | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Formulation | Range | Storage Requirements | Price | Quantity & Availability | ||
BMA50550
|
Lonza™
50550 |
500 μL | Markers have been labeled with 3 different Fluorescent dyes, Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load | 10 to 190 kd | −20°C |
Each for $459.00
|
|
||
BMA00193837
|
Lonza™
00193837 |
500 μL | Prestained using four different dyes, Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250 and 300kd | 4.6 to 300 kd | −20° to 4°C |
Each for $518.50
|
|
||
BMA50552
|
Lonza™
50552 |
100 μL | Markers have been labeled with 3 different Fluorescent dyes, Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load | 10 to 190 kd | −20°C |
Each for $148.80
|
|
||
Description
- Useful for monitoring the efficiency of separation during electrophoresis
- Confirm that transfer has occurred from gel to membrane in Western blotting
ProSieve Color Protein Marker
- Proteins in the marker have been labeled with 3 different fluorescent dyes
- Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load.
- Proteins have approximate masses of 10, 15, 20, 25, 40, 50, 80, 125, and 190kDa
- Each lot is individually calibrated against a ProSieve Unstained Protein Marker
ProSieve QuadColor Protein Marker
- Proteins are individually prestained using four different dyes, producing a brightly colored ladder with an easy-to-remember pattern
- Marker is supplied ready-to-use, no heating, further dilution or addition of a reducing agent is required before use
- Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250, and 300kDa
- Apparent molecular weights are provided for each lot
Specifications
Monitoring protein separation prior to staining and provide confirmation of protein transfer in Western blotting |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title