Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ BioWhittaker™ ProDoma Hybridoma Media
Specifications
Content And Storage | 2° to 8°C |
---|---|
Without Additives | No Glutamine, No Phenol Red |
Quantity | 1 L |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
BWBE020029Q
|
Lonza™
BE02029Q |
ProDoma 1 | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
BWBE02032Q
|
Lonza™
BE02032Q |
ProDoma 3 | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
The original hybridoma media for murine, human, and chimeric origin cells.
- Non-animal, protein-free media for your hybridoma needs.
- Scalable from small flasks to large-scale bioreactors.
- Hybridoma cell growth
- Monoclonal antibody production
Specifications
2° to 8°C | |
No Glutamine, No Phenol Red | |
1 L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title