Learn More
Lonza™ BioWhittaker™ ProDoma Hybridoma Media
Supplier: Lonza™ BE02032Q
Description
The original hybridoma media for murine, human, and chimeric origin cells.
- Non-animal, protein-free media for your hybridoma needs.
- Scalable from small flasks to large-scale bioreactors.
- Hybridoma cell growth
- Monoclonal antibody production
Specifications
2° to 8°C | |
ProDoma 3 | |
No Glutamine, No Phenol Red | |
1 L |