Learn More
Lonza™ BioWhittaker™ Hybridoma Serum-Free Culture Media: HL-1 Complete
Prescreened in actual hybridoma systems to ensure minimal variation
Supplier: Lonza™ 77201
This item is not returnable.
View return policy
Description
- HL-1 supports the serum-free growth of hybridomas and differentiated cells of lymphoid origin
Specifications
2° to 8°C | |
Liquid | |
No Glutamine | |
2 × 500mL |