Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Long Crucible Tongs with Corrugated Tips, 20 in.
Fisherbrand™ Crucible Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.
Description
The long design facilitates use when reaching into extreme hot or cold environments. These tongs feature integral thumb and finger openings.
- Long-neck serrated tips
- Raised riveted joing
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Certifications
ISO 9001-02, GMP, cGMP
Specifications
1 | |
Stainless Steel | |
Crucibles | |
20 in. |
508 mm | |
Crucible Tongs | |
Tongs |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction