Learn More
ShockWatch™ LOG•IC Single Use Temperature Recorder
SDP
Designed to help optimize the cold chain by alerting manufacturers, handlers and shippers when a product has been exposed to temperature conditions beyond a specified threshold. ShockWatch LOG•IC Single Use Temperature Recorders are water resistant (NEMA 4) recorders with USB in addition to integrated wireless capability that allows for fast data downloads.
Supplier:  ShockWatch™ SWIPMT8
Encompass
 View more versions of this product
Catalog No. 22-111-021
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
Description
Description
  • Captures data every 60 seconds
  • Up to eight temperature alarm thresholds
  • Validated and FDA 21 CFR Part 11-compliant reports
  • Up to 255 uses per device 
  • Available with Stainless and Ribbon Probes
Specifications
Specifications
Three-year run life
-22°F to +167°F
Multi-Use Temperature Recorder (26 uses)
4000 logged data points / 16 million histogram data points
66
Product Suggestions
Product Suggestions