Learn More
Eppendorf™ LoBind Microcentrifuge Tubes: Protein
Best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus sample
Supplier: Eppendorf™ 925000090
Description
When biological samples are stored or incubated in standard reaction vessels more than 90% sample material can be lost within 24 hours due to adsorption to the plastic surface. Eppendorf LoBind Tubes and Plates maximize sample recovery by significantly reducing sample-to-surface binding – without the use of any type of coating. A combination of specially developed technology and ultrapure selected polypropylene ensures best recovery rates of your valuable samples.
Features and benefits
- LoBind material guarantees maximum sample recovery for improved assay results
- Free of surface coating (e. g. siliconization). Eliminates the risk of sample interference
- Available in tube and plate formats for easy up-scaling
- High stability for use in all sample preparation protocols
- PCR clean quality batch tested and certified by an independent laboratory for reliable purity
Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
Specifications
30,000 x g | |
1.5 mL | |
Polypropylene | |
400 | |
Safe-lock Lid |
DNA, DNase, RNase, PCR Inhibitors free | |
Clear | |
Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples | |
Non-sterile | |
Conical |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.