Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium Dodecyl Sulfate (Powder or Flakes/Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
Description
Lithium Dodecyl Sulfate (LDS) is a detergent used for dissociating proteins.
Specifications
Absorbance | 0.01 max. (3% solution) at 405nm, 0.05 max. (3% solution) at 280nm, 0.100 AU max. at 260nm, 0.3 max. (3% solution) at 230nm |
Color | White |
Physical Form | Solid |
Assay Percent Range | ≥99 % |
Linear Formula | CH3(CH2)11OSO3Li |
Quantity | 25 g |
Identification | Pass Test |
Formula Weight | 273.18 |
Percent Purity | ≥99% (≥98% C12-OH) |
Grade | Molecular Biology |
Show More |
Safety and Handling
Recommended Storage | RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction