Learn More
MetaSystems Liquillizer™ Mucolytic Agent

Liquillizer™ Mucolytic Reagent is ideal for fast, efficient, user friendly preparation of clinical sputum and other respiratory samples for microbiology or mycobacteriology testing.
Supplier: MetaSystems P7510001IG
Description
- Ready to Use, No Preparation or Mixing
- Odorless and DTT Free
- Room Temprature Storage
- No Incubation, Centrifuging, or Sample Heating
- 4 Week Open Container Shelf Life
Specifications
Non-sterile | |
1 Ea. | |
125 mL |
Diagnostic procedures and cultivation | |
Store and use at room temperature (15°C to 25°C) |
Safety and Handling
ShelfLife : 4 weeks after opening
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Liquillizer™ is intended for the liquefaction of viscous respiratory specimens such as sputa, tracheal secretions, bronchial secretions or bronchoalveolar lavage. The mucolytic Dithiothreitol (DTT) -free buffer reduces specimen viscosity for easier sample handling while maintaining organism viability. No interference with molecular methods has been observed