Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Liquid Scintillation Vials, PET
Provides the clarity of glass with the safety of plastic.
Supplier: DWK Life Sciences 986754
Description
Provides the clarity of glass with the safety of plastic.
- Vials offer low permeability to solvents and minimal background counts
- Clarity of glass with the safety of plastic
- Vials can be safely incinerated
- No harmful gas is generated
- Packaged in 5 utility trays with each partition tray holding 100 vials
- Utility trays serve as a way to store your samples
Specifications
Linerless | |
27 mm | |
20 mL | |
Polyethylene | |
22-400 |
Clear | |
PET | |
500 | |
61 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction