Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Liquid Chromatography Column, Open-Ended
Constructed of borosilicate glass and rated at a maximum pressure of 15 psi (1 bar)
Supplier: MilliporeSigma™ Supelco™ C64191EA
Description
- Fixed polypropylene ends (outside of tube only)
- Without jacket
- Without end fittings
- Requires 2 flow adapters
Specifications
1 Ea. | |
LPLC | |
Open-Ended | |
30 cm |
1.5 cm | |
Bed Volume: 53 mL | |
15 psi (1 bar) | |
Borosilicate Glass |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title