Learn More
Thermo Scientific™ Lindberg/Blue M™ LGO Box Furnace PROMO
LGO Box Furnaces feature the latest technical advances in heating elements, insulation and temperature control, all integrated into a self-contained cabinet.
Supplier: Thermo Scientific™ BF51842C1PM
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Thermo Scientific™ LGO Box Furnaces feature the latest technical advances in heating elements, insulation and temperature control, all integrated into a self-contained cabinet. Furnaces feature proprietary LGO (light gauge overbend) heating elements and Moldatherm™ insulation for efficient and economical transfer of heat to chamber, with low exterior temperatures.
- Variable heat-up rate eliminates thermal shock to materials with quick heat-up and cool-down rates
- Choice of side hinge or vertical lift door
- Air vent (1 in. dia., top) and air inlet (0.375 in. dia., rear) for inert atmosphere exchange (may experience some leakage at door)
- Self-tuning, digital instrumentation for precise temperature setpoint and display
- Platinel II™ thermocouple for long-term stability
- 0.6 cu. ft. models feature vertical lift door; 2 cu. ft. models feature horizontal side swing door, hot side facing away from operator for protection
Microprocessor Control
- Microprocessor-based self-tuning PID control (proportional, integral, derivative) provides optimum thermal process, prevents overshoot
- Control panel designed for easy access and maintenance
- Main power ON/OFF switch on control panel
- Adjustable high-limit over temperature protection
- Simultaneous LED display of actual temperature vs. setpoint
- Can be configured to display temperature in either °C or °F
- Safety door switch interrupts power to heating element when door is opened; protects heating elements and minimizes exposure to end-user
- Removable shelves for versatility
- Moldatherm™ hearthplate supports load and prevents damage due to spillage
Digital, Single-Setpoint Controller
- Single segment, single setpoint, one ramp to setpoint
Digital Single-Program, Multiple-Segment Programmable Controller
- Single program with multiple segments for ramp (up and down) and dwell (timed hold) temperature control
Digital Multiple-Program, Multiple-Segment Programmable Controller
- Available on models with
P designation Multiple programs and segments for ramp (up and down) and dwell (timed hold) temperature control
over temperature Control (OTC)†
- Adjustable digital over temperature control, available on selected models with
B suffix designation
- Protects furnace and load in the event of primary control circuit failure
- Overrides main controller and shuts off power to furnace if high limit is reached
- Manual reset required for safety
- Operates via magnetic contacts through signal from independent thermocouple
Flowmeter Option (FM) (Inert Atmosphere Only)
- Available on selected models with
FM designation
- Gas flowmeter, adjustable, located on front control panel
- Adjustable flowrate, range 1.0 to 10.0 cu. ft./hr. standard
- Suitable for inert gas or airflow to chamber
- Allows fresh air exchange for ashing applications Not suitable for combustible or volatile gases
Ordering Information:
Required power cord and hardwiring not included. Optional RS485 Digital Communications Port allows controller to be connected to a PC for remote monitoring and control of the furnace; up to 30 units can be connected to one PC.
†Specify Option
B for over temperature Control (OTC) when ordering.
Includes:
One two-part shelf (0.6 cu. ft. models have one shelf position at center position; 2.0 cu. ft. models have three shelf positions)
Specifications
2.0 cu. ft. | |
100°C to 1200°C | |
5800 W | |
38.1 x 38.1 x 38.1 cm (15 x 15 x 15 in.) | |
28 in. | |
29 in. | |
38.1 cm | |
15 in. | |
38.1 cm | |
38.1 cm | |
208/240 V 50/60 Hz | |
50/60 Hz |
55.3 L | |
Multisegment Muffle Furnace | |
Digital Mutli Seg/1 Prog | |
71.1 x 73.7 x 83.8 cm (28 x 29 x 33 in.) | |
71.1 cm | |
73.7 cm | |
33 in. | |
83.8 cm | |
15 in. | |
127 kg | |
208/240 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.