Learn More
Lightning Leaper
Watch electricity leap.
Supplier: Science First 6153165
Requires : Van de Graaff generator
Description
An insulating plate with two binding clips on either end connects to a Van de Graaff generator. On the plate's surface, an incomplete metallic path with eight small gaps is drawn in a zig-zag pattern. The discharge from the Van de Graaff follows the metallic path rather than the shortest path.
- Can be suspended from an insulating stand and connected at either end to a Van de Graaff generator
- Can be held in the hand close to the active Van de Graaff generator
- Results amplified in a darkened room
Specifications
25,000 to 30,000 V | |
Use to watch high voltage electricity jumping across the gaps in the plate in the form of sparks | |
Van de Graaff generator | |
Lightning Leaper Demonstration Aid |