Learn More
GENIE GROUP EasyView Light Meter EA30
Supplier: GENIE GROUP EA30
The EA30 measures 400,000Fc in 4 ranges and 400,000Lux in 5 ranges. It has a rugged design and features a large display. The data hold feature freezes the reading on the display. It has cosine and color corrected measurements.The EA30 comes compete with built-in stand, remote sensor with protective cover, protective rubbber holster, and 6 AAA batteries.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.