Learn More
LIFE™ CPR Mask with One-Way Valve
Offers protection at half the size of all leading pocket masks. LIFE™ CPR Masks have a universal fit and superior seal.
$194.70 - $287.76
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
19-097-426
![]() |
LIFE™
LIFE100 |
Mask and valve only |
Case of 20 for $194.70
|
|
19-097-425
![]() |
LIFE™
LIFE100B |
Mask, Valve, and Nylon zip-bag with belt-loop |
Case of 20 for $287.76
|
|
Description
- Invertible
- Universally fits both adults and children
- Superior seal of contoured soft medical PVC to conform to all faces
- One-way valve and filter provide hygienic protection which eliminates transmission of vomitous and contagious diseases
- Hydrophobic filter for maximum protection placed above the valve to prevent fluid contamination from victim
- Large airway opening prevents back-pressure
- Clear mask and valve allow excellent observation of their interiors, and victim's color and status
Specifications
CPR Mask |