Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LevGo smartSpatula™ Disposable Polypropylene Spatula, Opaque
SCI_ED

smartSpatulas offer an economical, disposable tool for transferring, collecting, and handling chemical and biological materials

$30.75 - $38.00

Specifications

Autoclavable Autoclavable
Color Opaque
For Use With (Application) Use as a spatula, spoon, scoop, scraper, stirring rod, or policeman
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Outer Price Quantity & Availability  
S28421
SCI_ED
View Documents
Levgo Inc
17231/EMD
140 mm 3.5mm
Pack of 300 for $30.75
Case of 25 PK for $695.00
Add to Cart
 
S28423
SCI_ED
View Documents
Levgo Inc
17221/EMD
210 mm 7mm
Pack of 300 for $38.00
Case of 25 PK for $861.00
Add to Cart
 
Description

Description

  • Economical, Polypropylene, Single-Use Laboratory Spatulas
  • Ideal for Transferring, Manipulating, and Mixing Materials
  • Versatile Double-Ended Design
  • Help Minimize Cross Contamination
  • Resistant to acids and bases
  • Autoclavable
Specifications

Specifications

Autoclavable
Opaque
Use as a spatula, spoon, scoop, scraper, stirring rod, or policeman
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit