Learn More
Verichem Laboratories LEVEL G ENZYME
Supplier: Verichem Laboratories 9417
Enzynme Er Verifier, Level G. Protein Based, Liquid, Ready To Use. Inteded To Text The Rang For Calibration Verification Of Au Chemistry Ast Only. Stable 14 Months From Date Of Manufacture. Specifically Designed For Beckman Au Chemistry Systems. Assy Value Included For All Au Systems.