Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead (Foil/Laboratory), Fisher Chemical

Catalog Number L271RL
Click to view available options
Quantity:
1 lb.
Packaging:
Roll
7439-92-1
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
5352425
CHEBI:27889
lead
[Pb]
7439-92-1
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
5352425
CHEBI:27889
lead
[Pb]

12“ wide, 8/1000“ thick (305cm, 02mm)

TRUSTED_SUSTAINABILITY
Color Blue
Melting Point 327.4°C
Boiling Point 1740°C
Density 11.3g/cm³
Quantity 1 lb.
Identification Pass Test
Formula Weight 207.2
Grade Laboratory
Packaging Roll
Vapor Pressure 1.3mmHg at 970°C
Form Solid
Chemical Name or Material Lead
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit