Learn More
Andwin Scientific LEAD DONUT VINYL COATED 454G
Supplier: Andwin Scientific EF8839B
The lead ring is vinyl-coated and in a shape of a donut. It can work with flasks with volume capacity of 250 to 1000 mL. This product allows flask to be immersed in water baths, lessening risks of breakage and spills. The lead ring weighs 454 grams.