Learn More
Andwin Scientific LEAD DONUT CSHAPE 1000-4000ML
Supplier: Andwin Scientific EF8880D
The Heathrow Scientific Vinyl-Coated Lead Rings feature resistance against chemicals. This product is made to perform as weights to keep flasks from moving or floating around during submersion in water baths. The C-Shaped lead rings are compatible with 1000 to 4000 mL Erlenmeyer Flasks.