Learn More
Lanx Chemical Protective Integrated Gloves
Catalog Number p-2715368
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Butyl rubber with TuffStuff leather palm for a more secure grip.
  • Cotton liner
  • Butyl rubber with leather palm
  • Ideal for chemical and biological applications
  • Vacuum packed
Color Black
Size Small